Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
SDP
Catalog No. K9538250000 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
1000 mL (Funnel), 4000 mL (Flask)
300 mL (Funnel), 1000 mL (Flask)
500 mL (Funnel), 2000 mL (Flask)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
K953825-0000 300 mL (Funnel), 1000 mL (Flask)
K953835-0000 500 mL (Funnel), 2000 mL (Flask)
K953845-0000 1000 mL (Funnel), 4000 mL (Flask)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K953825-0000 Supplier DWK Life Sciences Supplier No. 9538250000
Add to Cart
Add to Cart
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
  • The ground joint connection eliminates the phthalate contamination that can occur when using silicone or neoprene stoppers
  • The support base has a 40-60 micron porosity glass frit and an integral vacuum connection that is located above the drip tip to prevent contamination of the vacuum line with filtrate droplets
  • Each apparatus is supplied with a funnel, an anodized aluminum clamp, a 47 mm fritted glass support base and a filtration flask
  • Joints are 40/35
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Type Microfiltration Assembly
Volume (Metric) 300 mL (Funnel), 1000 mL (Flask)