Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column Stainless Steel Nut for 1/8 in. Tubing
SDP
Catalog No. K4208216218 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
K4208216218 Stainless Steel
1 options
Catalog No. K4208216218 Supplier DWK Life Sciences Supplier No. 4208216218
Add to Cart
Add to Cart
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™ and DISPOSAFLEX™ columns.
  • Stainless Steel Nut for 1/8 in. OD Tubing
  • 1/4”-28 Thread
  • Use with the 420822-Series Flangeless Ferrules
  • Case of 1
TRUSTED_SUSTAINABILITY
Specifications
Type Tubing Nut
Diameter (English) Outer 0.125 in.
Diameter (Metric) Outer 0.32 cm
Material Stainless Steel