Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ Kontes™ Variable Reflux Distillation Heads with PTFE Plugs
SDP

Catalog No. K2874730000 Shop All DWK Life Sciences Products
Change view
Click to view available options
:
Variable Reflux with PTFE Plug, 14/20 Joints
Variable Reflux with PTFE Plug, 19/22 Joints
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
K2874730000 Variable Reflux with PTFE Plug, 14/20 Joints
K2874731922 Variable Reflux with PTFE Plug, 19/22 Joints
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K2874730000 Supplier Kimble™ Supplier No. 2874730000

Lower receiver joint with drip tip

  • Rotation of cold finger condenser allows collection of distillate or return to flask
  • Stopcocks allow removal of the product or a receiver change without interrupting distillation
  • standard taper 10/30 outer side joint accepts 75mm immersion thermometer
  • Column standard taper joints: 24/40
  • Condenser standard taper joints: 29/42
  • Receiver standard taper joints: 24/40
  • Hose connection: #2
  • H x W: 420 x 200mm
TRUSTED_SUSTAINABILITY
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.