Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KOVA™ Refractrol SP

Catalog No. 22749706 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Description:
High Level
Low Level
Normal Level
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
22-749-706 High Level
22-749-704 Low Level
22-749-705 Normal Level
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-749-706 Supplier KOVA™ Supplier No. K84654
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KOVA Refractrol SP tri-level control provides 5 levels of serum protein control solutions for use in refractometry

  • For use with temperature compensated and non-compensated refractometers
  • Recovery of the assigned values provides assurance of measurements of plasma or serum protein concentration
  • Each level of Refractrol is separately packaged in easy-to-use 2mL vials
Storage: Store at room temperature (18 - 30°C). Do not refrigerate or freeze. Crystals or debris may be observed in the stain, this is not uncommon for Sternheimer-Malbin stain. These artifacts may be safely removed by filtration. Stable per the expiration date on the label.

Specifications

Description High Level
Quantity 12 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.