Learn More
Cytiva (Formerly Pall Lab) KPC HT MALE CONN 1/2IN 25PK
Supplier:  Cytiva KPCHT02M6
Encompass
The disposable Male Kleenpak sterile connector allows for the dry connection of two separate fluid pathways, while maintaining the sterile integrity of both Standard pricing not available. Cytiva requires a new quote with updated pricing for each order.
Catalog No. NC0162627
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.