Learn More
Selleck Chemical LLC KPT-185 10mg 1333151-73-7
SDP
Supplier:  Selleck Chemical LLC S712510MG
Encompass_Preferred
KPT-185 is a selective CRM1 inhibitor that induces growth inhibition and apoptosis in a panel of NHL cell lines with a median IC50 ~25 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0785740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.