Learn More
CPC Scientific H-Lys-Arg-Ile-Val-Gln-Arg-Ile-Lys-Asp-Phe-Leu-Arg-NH2 5MG
SDP
Supplier:  CPC Scientific LL37001B
Encompass
KR-12 corresponds to amino acid residues 18-29 of LL-37 and is the smallest peptide of LL-37 retaining antibacterial activity. KR-12 displays a selective toxic effect on bacteria but not on human cells. It may be used as a template for developing novel antimicrobial agents of therapeutic use.SEQUENCE: H-Lys-Arg-Ile-Val-Gln-Arg-Ile-Lys-Asp-Phe-Leu-Arg-NH2ONE-LETTER SEQUENCE: KRIVQRIKDFLR-NH2MOLECULAR FORMULA: C71H127N25O15MOLECULAR WEIGHT: 1570.95STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [1218951-51-9]SYNONYMS: Antibacterial Protein LL-37 amide (human) (18-29), LL-37 (18-29), LL37 (18-29), CAMP (18-29), KR-12 amideRESEARCH AREA: AntimicrobialREFERENCES:G.Wang, J. Biol. Chem., 283, 32637 (2008)SKU(s): LL37-001A, LL37-001B, LL37-001C
Catalog No. 50-210-9680
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.