Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abrasive Sheets, Krackeler Scientific
SDP

Catalog No. 50615896N
Change view
Click to view available options
Chemical Name or Material:
Abrasive sheet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
50615896N Abrasive sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50615896N Supplier KSE Scientific Supplier No. 9250615896EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sheets, Abrasive, Krackeler

Specifications

Physical Form Sheet
Chemical Name or Material Abrasive sheet
Packaging Abrasive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.