Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Krackeler Scientific 4mL Screw Top Vials, Caps and Septa
SDP

Catalog No. 0767340180N
Change view
Click to view available options
Product Type:
Diffusion Insert
Wash Vial
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
0767340180N Diffusion Insert
51820551N Wash Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0767340180N Supplier Krackeler Scientific Supplier No. 920767340180EA
Add to Cart
Add to Cart

Specifications

Color Black
Closure Type Open Top Cap
For Use With (Equipment) 4 mL Screw Thread Vials
Product Type Diffusion Insert
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.