Learn More
Krackeler Scientific Replacement Needles
SDP
Catalog No. 51820836N
Change view
Click to view available options
Volume (Metric):
5 μL
Catalog No. Volume (Metric)
51820836N 5 μL
1 options
Catalog No. 51820836N Supplier Krackeler Scientific Supplier No. 9251820836EA
Specifications
Diameter (Metric) Inner Needle 0.25 to 0.53 mm
For Use With (Application) Autosampling
Product Type Replacement Needle
Quantity Ea.
Volume (Metric) 5 μL
For Use With (Equipment) 7673/7683 On-column Autosampler Syringe