Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRAP Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501568174
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8174 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8174 Supplier Bethyl Laboratories, Inc Supplier No. A304607AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

SSFA2 gene ontology annotations related to this gene include actin binding.

Specifications

Antigen KRAP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SSFA2
Gene Accession No. P28290
Gene Alias CS-1, CS1, KRAP, SPAG13, KRAS-induced actin-interacting protein, cleavage signal-1 protein, ki-ras-induced actin-interacting protein, sperm associated antigen 13, sperm-specific antigen 2
Gene Symbols SSFA2
Host Species Rabbit
Immunogen Between 1209 and 1259
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6744
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.