Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Krebs-Ringer Solution, without calcium

Catalog No. AAJ67839K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67839K2 1 L
AAJ67839AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67839K2 Supplier Thermo Scientific Chemicals Supplier No. J67839K2
  • $224.50
    $190.65 / Each of 1

    Save $33.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Quantity 1 L
Packaging Plastic Bottle
What is the pH of the Krebs-Ringer Solution, without calcium?

The pH of the Krebs-Ringer Solution, without calcium is 7.3.

What are the concentrations of the components of Krebs-Ringer Solution, without calcium?

The Krebs-Ringer Solution, without calcium contains 120 mM sodium chloride, 5 mM potassium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, and 5.5 mM D-glucose.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.