Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Kremen1 (Mouse) ELISA Kit

Catalog No. 89335212 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-212 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-212 Supplier Abnova™ Supplier No. KA5176
Add to Cart
Add to Cart

Sandwich enzyme-linked immunosorbent assay

Kremen1 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of mouse Kremen1 in cell culture supernates, cell lysates, serum and plasma (heparin, EDTA).

Specifications

Description kringle containing transmembrane protein 1
Cross Reactivity Mouse
Target Species Mouse
For Use With (Application) Quantitative
Gene ID (Entrez) 84035
Quantity 1 Kit
Regulatory Status RUO
Storage Requirements Store at 4°C for six months. For long term storage store at -20°C. Avoid repeated freezing and thawing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.