Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KREMEN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534520
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534520 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534520 Supplier Invitrogen™ Supplier No. PA534520
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat small intestine tissue lysate. PA5-34520 can be used with blocking peptide PEP-1563.

KREMEN1 is a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. This protein is a component of a membrane complex that modulates canonical WNT signaling through lipoprotein receptor-related protein 6 (LRP6). It contains extracellular kringle, WSC, and CUB domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KREMEN1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Kremen1
Gene Accession No. Q924S4, Q96MU8, Q99N43
Gene Alias AV002070; dickkopf receptor; KREMEN; kremen protein 1; KREMEN1; kringle containing transmembrane protein 1; kringle domain-containing transmembrane protein 1; kringle-coding gene marking the eye and the nose; kringle-containing protein marking the eye and the nose; KRM1
Gene Symbols Kremen1
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human KREMEN1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114107, 83999, 84035
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.