Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the quantitative measurement of E.coli Host Cell Proteins as a contaminant in various biological preparations. Assay Range: 3.125 - 200 ng/ml; Sensitivity: 1.875 ng/ml.
Supplier:  KRISHGEN BIOSYSTEMS KBBP01
Encompass_Preferred
Pack Size: 1 x 96 wells. Introduction: A variety of proteins and pDNA which are used as therapeutic agents in humans and animals are produced through recombinant expression in E.coli. The manufacturing and purification process of these products tends to leave the potential for contamination by Host Cell Proteins (HCPs) from E.coli which may result in adverse toxic or immunological reactions that ultimately affect the efficacy of the therapeutic agent. The simple, objective and semi-quantitative ELISA is a highly sensitive method that aids in purification process development, process control, quality control and product release testing optimally. This E.coli HCP ELISA kit offers accurate, quick results and is validated for lot-to-lot consistency. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Precision CV <10%.
Catalog No. NC2747134
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.