Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT75 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553987
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553987 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553987 Supplier Invitrogen™ Supplier No. PA553987
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSRQSSITFQ SGSRRGFSTT SAITPAAGRS RFSSVSVARS AAGSGGLGRI SSA Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 79%.

This gene is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT75
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT75
Gene Accession No. O95678
Gene Alias 4732468K03Rik; AA589387; CK-75; cytokeratin KRT2-6HF; cytokeratin type II; cytokeratin-75; hK6hf; K6HF; K75; KB18; keratin 75; keratin 75, type II; keratin, type II cytoskeletal 75; Keratin-6 hair follicle; keratin-75; keratinocyte associated protein 1; Krt2-6hf; KRT75; Krtcap1; mK6hf; PFB; type II keratin Kb18; type II keratin-18; type II keratin-K6hf; Type-II keratin Kb18
Gene Symbols KRT75
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRT75. Recombinant protein control fragment (Product #RP-92529).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9119
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.