Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRTAP27-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561821 Supplier Thermo Scientific Supplier No. PA561821
Only null left
Add to Cart
Add to Cart

KRTAP27-1 Polyclonal Antibody

KRTAP27-1 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen KRTAP27-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRTAP27-1
Gene Accession No. Q3LI81
Gene Alias KAP27.1; keratin associated protein 27-1; keratin-associated protein 27-1; KRTAP27-1
Gene Symbols KRTAP27-1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRTAP27-1. Recombinant protein control fragment (Product #RP-91805).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643812
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.