Learn More
MAPA™ Krytech 576 Cut-Resistant A4 Polyurethane Coated Glove
Catalog No. 19002841 Shop All MAPA Professional Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-002-841 6
19-002-842 7
19-002-843 8
19-002-844 9
19-002-845 10
19-002-846 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. 19-002-841 Supplier MAPA™ Supplier No. 34576036
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Krytech 576 Cut-Resistant A4 Polyurethane Coated Glove - Optimal Cut Resistance. Excellent Dexterity due to precise fit. Added Comfort and Flexibility. HDPE Liner.
  • Automotive Industry
  • Handling Metal Parts
  • Assembly of Thin Metal Sheets
  • Finishing of Sharp Parts
  • Plastics Industry
  • Post Injection Material Removal
  • Construction Industry
  • Roofers
  • Window Installers
  • Tilers
  • Glazers
Specifications
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material HDPE
Product Type Gloves
Size 6
For Use With (Application) Automotive Industry, Handling Metal Parts, Assembly of Thin Metal Sheets, Finishing of Sharp Parts, Plastics Industry, Post Injection Material Removal, Construction Industry, Roofers, Window Installers, Tilers, Glazers
Gender Unisex
Glove Cuff Style Knit
Puncture Rating ASTM F2878 LEVEL 0
Series Krytech 576
Thumb Style Straight
Yarn Gauge 13