Learn More
Apexbio Technology LLC KT 5720 108068-98-0 500ug
Supplier: Apexbio Technology LLC B90020.5
KT 5720 (CAS 108068-98-0) is a cell-permeable reversible inhibitor of protein kinase A (PKA) that functions through ATP-competitive antagonism with an experimentally determined IC50 of 3 3 M This compound has been shown to counteract multidrug resistance in tumor cells mediated by MDR1 Additionally KT 5720 suppresses hyperpolarization-activated cyclic nucleotide-gated (HCN) cation channel activity resulting in decreased intracellular calcium levels and reduced excitability in dorsal root ganglion (DRG) neurons KT 5720 is utilized in studies investigating mechanisms underlying hematological malignancies and HCN or DRG-related disorders
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.