Learn More
Apexbio Technology LLC U 73343 142878-12-4 25mg
SDP
Supplier:  Apexbio Technology LLC B901325
Encompass_Preferred
U 73343 (CAS 142878-12-4) is a structural analog of U 73122 commonly utilized as an inactive control in studies investigating phospholipase C (PLC) activity Unlike U 73122 U 73343 does not inhibit PLC-mediated hydrolysis of phosphatidylinositol 4 5-bisphosphate a key step generating diacylglycerol and inositol 1 4 5-trisphosphate involved in intracellular signaling Through its lack of PLC antagonism U 73343 serves as a negative control to distinguish nonspecific or off-target effects in experiments analyzing processes such as hormone-induced calcium mobilization and gastric acid secretion thereby supporting accurate assessment of PLC-dependent signaling mechanisms
Catalog No. 50-000-89297
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.