Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab KT5i+ Premium German Advanced Adhesion InkJet Compatible Slides
SDP

Catalog No. 21402408
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Corner Style:
90°
90° Clipped
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Corner Style
21-402-408 Yellow 90°
21-402-400 Blue 90°
21-402-401 Blue 90° Clipped
21-402-402 Green 90°
21-402-403 Green 90° Clipped
21-402-404 Pink 90°
21-402-405 Pink 90° Clipped
21-402-406 White 90°
21-402-407 White 90° Clipped
21-402-409 Yellow 90° Clipped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 21-402-408 Supplier StatLab Supplier No. KT5PLUSIYL90
Add to Cart
Add to Cart

KT5+i Premium Advanced Adhesion InkJet Slides are compatible with both InkJet and UV laser printers

  • An excellent choice for H and E, special stains, frozen sections and even the toughest IHC procedures with difficult tissues
  • These slides feature superior adhesion, maintaining tissue integrity throughout the toughest of tissues or protocols
  • Tissue sections easily jump on the slide, indicating a hydrophobic waterbath behavior, but the precision manufacturing process ensures a 100% wettable surface for even reagent spreading
  • KT5+i stained slides will not have background staining
  • KT5+i slides provide a colored marking area for efficient specimen color-coding, identification, labeling and printing
  • KT5+i slides are available in 5 unique colors, with 45° or 90° corners
  • KT5+i Slides are compatible with both inkjet and UV laser printers
  • Manufactured from premium German glass that enables superior clarity for digital pathology and imaging systems
  • Dimenstions: 75.6 x 25.6 x 1 mm
  • 10 gross per case.

Specifications

Length (Metric) 75.6 mm
Frosted 1 End, 1 Side
Thickness (Metric) 1 mm
Well Count 0
Edges 90°, Ground
Sterility Non-sterile
Precleaned Yes
Material High Quality Glass
Width (Metric) 25.6 mm
Type Adhesion Slide
Color Yellow
Corner Style 90°
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.