Learn More
Hach Company HQ1140 Portable Dedicated Conductivity/TDS Meter with Conductivity Electrode, 5 m Rugged Cable
Supplier:  Hach Company LEV015.53.11402
Encompass
HQ1140 Portable EC/TDS Meter, 1 channel for general-purpose field or plant testing with CDC401 laboratory 4-poles graphite conductivity cell, 5 m rugged cable. Field accessories included.
Catalog No. 50-206-2944