Learn More
Hach Company HQ2100 Portable Multi-Meter with Conductivity Electrode, 1 m Cable
Supplier:  Hach Company LEV015.53.21006
Encompass_Preferred
HQ2100 Portable pH/ORP/EC/TDS/DO Meter, 1 channel for general-purpose field or plant testing with IntelliCAL CDC401 laboratory luminescent/optical dissolved oxygen electrode, 1 m cable. Field accessories included.
Catalog No. 50-206-2946
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.