Learn More
Enzo Life Sciences KU55933 (5mg). CAS: 587871-26-9
Supplier: Enzo Life Sciences ENZCHM1500005
KU55933 is a potent and selective ATP-competitive inhibitor of ATM, with an IC50 of 13 nmol/l and a Ki of 2.2 nmol/l. KU55933 has potential for use as a new radio- and chemo-sensitizing agent for the treatment of cancer. Alternative name: ATM kinase inhibitor, 2-Morpholino-6-(thianthren-1-yl)-4H-pyran-4-one. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (50 mg/ml) or ethanol (<13 mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.