Learn More
Invitrogen™ Ku80 Recombinant Rabbit Monoclonal Antibody (5G3)

Description
This antibody has been tested in direct-ELISA.
Specifications
Specifications
| Antigen | Ku80 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 5G3 |
| Concentration | 0.08 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 150 mM NaCl and 0.02% sodium azide; pH 7.4 |
| Gene | XRCC5 |
| Gene Accession No. | P13010 |
| Gene Alias | 86 kDa subunit of Ku antigen; AI314015; ATP-dependent DNA helicase 2 subunit 2; ATP-dependent DNA helicase II 80 kDa subunit; CTC box-binding factor 85 kDa subunit; CTC85; CTCBF; DNA repair protein XRCC5; FLJ39089; G22p2; HGNC:12833; KARP1; KARP-1; Ku autoantigen; ku autoantigen protein p86 homolog; Ku autoantigen, 80kD); Ku autoantigen, 80kDa; Ku p80; Ku80; Ku-80; Ku86; Ku-86; Ku86 autoantigen related protein 1; KUB2; Kup80; Lupu; lupus; lupus Ku autoantigen protein p86; NFIV; nuclear factor IV; OTTHUMP00000164061; OTTHUMP00000206791; Thyroid-lupus autoantigen; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 5; X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining; Ku autoantigen, 80kDa); X-ray repair cross complementation (double-strand-break rejoining; X-ray repair cross complementation (double-strand-break rejoining; Ku autoantigen, 80kD); X-ray repair cross complementing 5; X-ray repair cross-complementing protein 5; Xrcc5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.