Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Kut-Gard™ Polyester-Wrapped Stainless Steel Core Cut-Resistant Gloves

Catalog No. 191304477E Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-4477E X-Large
19-130-4477A X-Small
19-130-4477B Small
19-130-4477C Medium
19-130-4477D Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-130-4477E Supplier PIP™ Supplier No. 22650XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for any application where sharp cutting instruments are used such as food service, food processing, and glass and metal handling.

  • Seamless knit construction provides comfort, fit and dexterity
  • PolyKor™ engineered yarns offer cut and slash resistance
  • Ambidextrous pattern permits glove to be worn on either hand
  • Special non-latex cuff dramatically reduces cuff blowout, even after multiple launderings in bleach
  • Can be worn on its own or as a glove liner
  • Secure gripping properties
  • Shrink resistant
  • Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199
  • Products constructed with PolyKor™ engineered yarn are suitable for protection against mechanical hazards in the toughest environments
  • PolyKor products are highly resistant to abrasion and chemicals, so you can rely on them to provide long-lasting protection

Specifications

Certifications/Compliance FDA 21 CFR, 170-199
Construction Seamless Knit
Coating Material Uncoated
Color Blue
Cut Resistance Rating ANSI A5
Hand Ambidextrous
Latex Free Yes
Material Stainless Steel Fiber, Synthetic Fiber
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Application where sharp cutting instruments are used, such as food service, food processing and glass/metal handling
Glove Cuff Color Green
Glove Cuff Style Elasticated
Hem Color Green
Model 22-650XL
Series Kut-Gard™ 22-650
Yarn Gauge 7
Length (English) 11 in.
Length (Metric) 27.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.