Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ KuWal151

Catalog No. p-7166787
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-441-0 10 mg
66-445-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 66-441-0 Supplier Tocris™ Supplier No. 6644/10
This item is not returnable. View return policy

Potent and selective CLK inhibitor

KuWal151 is a potent and selective CLK inhibitor (IC50 values are 28, 88 and 510 nM for CLK4, 1 and 2, respectively). Exhibits no significant activity at CLK3, DYRK, CDK or GSK3 at a concentration of 10 μM. Inhibits growth of a range of cancer cell lines in vitro at subnanomolar concentrations.

Specifications

Quantity 10 mg
Molecular Formula C16H11ClN2O
Formula Weight 282.72
Product Type Inhibitor
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.