Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KV1.5 (KCNA5) Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674811
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-811 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-811 Supplier Invitrogen™ Supplier No. PA112656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for this product is rat brain lysate.

In neurons voltage-dependent potassium channels are key determinants of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV1.5 (KCNA5)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene Kcna5
Gene Accession No. P19024, P22460, Q61762
Gene Alias ATFB7; cardiac potassium channel; delayed rectifier K+ channel; HCK1; HK2; HPCN1; insulinoma and islet potassium channel; KCNA5; Kv1; KV1.5; KV1-5; MGC117058; MGC117059; PCN1; Potassium (K+) channel protein alpha 5; potassium channel 1; potassium channel, voltage gated shaker related subfamily A, member 5; potassium voltage gated channel, shaker related subfamily, member 5; potassium voltage-gated channel subfamily A member 5; potassium voltage-gated channel, shaker-related subfamily, member 5; RCK7; RK4; voltage-gated potassium channel HK2; voltage-gated potassium channel protein Kv1.5; voltage-gated potassium channel subunit Kv1.5
Gene Symbols Kcna5
Host Species Rabbit
Immunogen Synthetic peptide, conjugated to KLH, corresponding to the rat Kv15 potassium channel.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16493, 25470, 3741
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.