Learn More
Invitrogen™ KV2.1 (KCNB1) Polyclonal Antibody

Description
KV2.1 (KCNB1) Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | KV2.1 (KCNB1) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | Kcnb1 |
| Gene Accession No. | P15387, Q03717, Q14721 |
| Gene Alias | Delayed rectifier potassium channel 1; delayed rectifier potassium channel Kv2.1; DRK1; DRK1PC; KCB1; Kcnb1; Kcr1-1; Kv2.1; mShab; potassium channel Kv2.1; potassium channel, voltage gated Shab-related subfamily B, member 1; potassium voltage gated channel, Shab-related subfamily, member 1; potassium voltage-gated channel subfamily B member 1; potassium voltage-gated channel, Shab-related subfamily, member 1; Shab; voltage-gated potassium channel subunit Kv2.1 |
| Gene Symbols | Kcnb1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.