Learn More
Description
This antibody detects endogenous protein at a molecular weight of 70 kDa. Purity is >95% by SDS-PAGE.
Specifications
Specifications
| Antigen | KV3.2 (KCNC2) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide; pH 7.2 |
| Gene | KCNC2 |
| Gene Accession No. | P22462, Q14B80, Q96PR1 |
| Gene Alias | AW047325; B230117I07; KCNC2; KShIIIA; Kv3.2; Potassium channel voltage-gated Shaw-related subfamily C member 2; potassium channel, voltage gated Shaw related subfamily C, member 2; potassium channel, voltage gated Shaw-related subfamily C, member 2; potassium voltage gated channel, Shaw-related subfamily, member 2; potassium voltage-gated channel subfamily C member 2; potassium voltage-gated channel, Shaw-related subfamily, member 2; shaw-like potassium channel; Voltage-gated potassium channel Kv3.2; voltage-gated potassium channel subunit Kv3.2 |
| Gene Symbols | KCNC2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
