Learn More
Invitrogen™ KV3.3 (KCNC3) Polyclonal Antibody

Description
Peptide sequence: YAERIGADPD DILGSNHTYF KNIPIGFWWA VVTMTTLGYG DMYPKTWSGM Sequence homology: Cow: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | KV3.3 (KCNC3) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | KCNC3 |
| Gene Accession No. | Q14003 |
| Gene Alias | KCNC3; Kcr2-3; KShIIID; KV3.3; LOW QUALITY PROTEIN: potassium voltage-gated channel subfamily C member 3; potassium channel, voltage gated Shaw related subfamily C, member 3; potassium channel, voltage gated Shaw-related subfamily C, member 3; potassium voltage gated channel, Shaw-related subfamily, member 3; potassium voltage-gated channel subfamily C member 3; potassium voltage-gated channel, Shaw-related subfamily, member 3; SCA13; Shaw type potassium channel Kv3.3; Shaw-related voltage-gated potassium channel protein 3; voltage-gated potassium channel protein KV3.3; voltage-gated potassium channel subunit Kv3.3 |
| Gene Symbols | KCNC3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.