Learn More
KV3.4 (KCNC4) Monoclonal Antibody (S72-16), Invitrogen™
Description
Specifications
Specifications
| Antigen | KV3.4 (KCNC4) |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | S72-16 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 |
| Gene | KCNC4 |
| Gene Accession No. | Q03721, Q63734, Q8R1C0 |
| Gene Alias | AI850292; C1orf30; HKSHIIIC; K+ channel subunit; KCNC4; Kcr2-4; KSHIIIC; Kv3.4; MGC126818; potassium channel, voltage gated Shaw related subfamily C, member 4; potassium channel, voltage gated Shaw-related subfamily C, member 4; potassium voltage gated channel, Shaw-related subfamily, member 4; potassium voltage-gated channel subfamily C member 4; potassium voltage-gated channel subfamily C member 4 (Voltage-gated potassium channel subunit Kv3.4) (Raw3); potassium voltage-gated channel, Shaw-related subfamily, member 4; raw3; Voltage-gated potassium channel subunit Kv3.4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.