Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KYL
GREENER_CHOICE

Catalog No. 52901
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-901 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 52-901 Supplier Tocris Bioscience™ Supplier No. 5290/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

EphA4 kinase inhibitor; neuroprotective

KYL is an EphA4 receptor tyrosine kinase inhibitor (Kd = 0.8 μM); inhibits EphA4-EphrinA5 interactions (IC50 = 6.34 μM). Prevents AβO induced synaptic damage, dendritic spine loss and prevents the blocking of LTP in hippocampal CA3-CA1 transmissions. Exhibits a long half life in cell culture media (8 and 12 hours in PC3 and C2C12 media respectively). Neuroprotective.

Specifications

Description KYL
Molecular Weight (g/mol) 1465.75
Product Type KYL
Quantity 1 mg
Sequence Lys-Tyr-Leu-Pro-Tyr-Trp-Pro-Val-Leu-Ser-Ser-Leu

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.