Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kynurenic Acid Hydrate 98.0+%, TCI America™
SDP

Catalog No. H03031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03031G 1 g
H03035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03031G Supplier TCI America Supplier No. H03031G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 492-27-3
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00006753
InChI Key HCZHHEIFKROPDY-UHFFFAOYSA-N
Synonym kynurenic acid, 4-hydroxyquinoline-2-carboxylic acid, kynurenate, quinurenic acid, transtorine, kinurenic acid, kynuronic acid, 4-oxo-1,4-dihydroquinoline-2-carboxylic acid, 4-hydroxyquinaldic acid, 4-hydroxy-2-quinolincarboxylic acid
PubChem CID 3845
ChEBI CHEBI:18344
IUPAC Name 4-oxo-1H-quinoline-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C=C(N2)C(=O)O

Specifications

Quantity 1 g
Formula Weight 189.17
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Kynurenic Acid Hydrate
RTECSNumber VB2080000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.