accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 492-27-3

IUPAC Name: 4-oxo-1,4-dihydroquinoline-2-carboxylic acid
Synonyms: acid, kynurenic 4-hydroxyquinaldinate kynuronic acid 4-hydroxyquinaldate 4-hydroxy-quinaldate kinurenic acid kynurenic acid (substance) quinurenic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: HCZHHEIFKROPDY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)C2=CC=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results