Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-165,041, Tocris Bioscience™

Catalog No. 185650
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-565-0 50 mg
18-561-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-565-0 Supplier R&D Systems Supplier No. 1856/50
This item is not returnable. View return policy

Potent PPARδ agonist

Potent PPARδ agonist (Ki = 6 nM); displays > 100-fold selectivity for both mouse and human PPARδ receptors over other subtypes. Raises plasma cholesterol levels in vivo, in insulin-resistant db/db mice. Neuroprotective in models of cerebral infarction and Parkinson's disease.

Chemical Identifiers

CAS 79558-09-1
Molecular Formula C22H26O7
Molecular Weight (g/mol) 402.443
InChI Key HBBVCKCCQCQCTJ-UHFFFAOYSA-N
Synonym 2-4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy acetic acid, 4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy-acetic acid, 4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxyacetic acid, 2-4-3-4-acetyl-3-hydroxy-2-propylphenoxy-propoxy phenoxy acetic acid, 4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy acetic acid, 4-3-2-propyl-3-hydroxy-4-acetyl phenoxy propyloxyphenoxy-acetic acid, aceticacid, 2-4-3-4-acetyl-3-hydroxy-2-propylphenoxy propoxy phenoxy, pubchem19280, lopac-l-2167
PubChem CID 6603901
IUPAC Name 2-[4-[3-(4-acetyl-3-hydroxy-2-propylphenoxy)propoxy]phenoxy]acetic acid
SMILES CCCC1=C(C=CC(=C1O)C(=O)C)OCCCOC2=CC=C(C=C2)OCC(=O)O

Specifications

Quantity 50 mg
Formula Weight 402.44
Percent Purity >98%
Chemical Name or Material L-165, 041
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.