Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-2-Allylglycine Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. A1648200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1648200MG 200 mg
A16481G 1 g
A16485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1648200MG Supplier TCI America Supplier No. A1648200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 195316-72-4
Molecular Formula C5H10ClNO2
Molecular Weight (g/mol) 151.59
MDL Number MFCD06797042
InChI Key DIDZZOWASZMNQW-WCCKRBBISA-N
Synonym s-2-aminopent-4-enoic acid hydrochloride, l-2-allylglycine hydrochloride, s---2-amino-4-pentenoic acid hydrochloride, 2s-2-aminopent-4-enoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, 2s, l-allylglycine hydrochloride, l-alpha-allylglycine hydrochloride, s-2-amino-4-pentenoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, s
PubChem CID 44630067
IUPAC Name (2S)-2-aminopent-4-enoic acid;hydrochloride
SMILES C=CCC(C(=O)O)N.Cl

Specifications

Quantity 200 mg
Formula Weight 151.59
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-2-Allylglycine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.