Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-364,373, Tocris Bioscience™

Catalog No. 266050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-605-0 50 mg
26-601-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-605-0 Supplier R&D Systems Supplier No. 2660/50
This item is not returnable. View return policy

KV7.1 channel activator, activates IKs

Activator of KV7.1 (KCNQ1) channels. Activates the cardiac IKs current that decreases action potential duration (APD) in cardiac myocytes.

Chemical Identifiers

CAS 103342-82-1
Molecular Formula C25H20FN3O
Molecular Weight (g/mol) 397.453
InChI Key CGBANSGENFERAT-JOCHJYFZSA-N
Synonym r-l3, cck antagonist synthetic 11, 3r-5-2-fluorophenyl-3-1h-indol-3-ylmethyl-1-methyl-3h-1,4-benzodiazepin-2-one, d05gqk, 5-2-fluorophenyl-1,3-dihydro-3-1h-indol-3-ylmethyl-1-methyl-2h-1,4-benzodiazepin-2-one
PubChem CID 656755
ChEBI CHEBI:34942
IUPAC Name (3R)-5-(2-fluorophenyl)-3-(1H-indol-3-ylmethyl)-1-methyl-3H-1,4-benzodiazepin-2-one
SMILES CN1C2=CC=CC=C2C(=NC(C1=O)CC3=CNC4=CC=CC=C43)C5=CC=CC=C5F

Specifications

Quantity 50 mg
Formula Weight 397.44
Percent Purity >98%
Chemical Name or Material L-364, 373
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.