Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-798,106
GSA_VAGREENER_CHOICE

Catalog No. 334210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-4210 10 mg
33-425-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-4210 Supplier Tocris Bioscience™ Supplier No. 3342/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and highly selective EP3 antagonist

Potent and highly selective prostanoid EP3 receptor antagonist (Ki values are 0.3, 916, > 5000 and > 5000 nM at EP3, EP4, EP1 and EP2 respectively). Attenuates sulprostone-induced inhibition of EFS-evoked twitch and contractile responses in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 244101-02-8
Molecular Formula C27H22BrNO4S

Specifications

Quantity 10 mg
Formula Weight 536.44
Purity >99%
Molecular Weight (g/mol) 536.44
Chemical Name or Material L-798, 106
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.