Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanine Benzyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A277125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A277125G 25 g
A27715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A277125G Supplier TCI America Supplier No. A277125G

Chemical Identifiers

CAS 5557-83-5
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.677
MDL Number MFCD00054340
InChI Key RLMHWGDKMJIEHH-QRPNPIFTSA-N
Synonym h-ala-obzl.hcl, l-alanine benzyl ester hydrochloride, h-ala-obzl hcl, s-benzyl 2-aminopropanoate hydrochloride, l-alanine benzyl ester hcl, benzyl l-alaninate hydrochloride, benzyl 2s-2-aminopropanoate hydrochloride, h-ala-obzlhcl, ala-obzl hcl, nh2-ala-obn hcl
PubChem CID 12210893
IUPAC Name benzyl (2S)-2-aminopropanoate;hydrochloride
SMILES CC(C(=O)OCC1=CC=CC=C1)N.Cl

Specifications

Melting Point 138°C
Color White
Quantity 25 g
Formula Weight 215.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Alanine Benzyl Ester Hydrochloride
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.