Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanine isopropyl ester hydrochloride, 95%, Thermo Scientific™

Catalog No. AAH6249003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6249003 1 g
AAH6249006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6249003 Supplier Thermo Scientific Chemicals Supplier No. H6249003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
L-Alanine isopropyl ester hydrochloride is a reagent used in the synthesis of anti- hepatitis C virus prodrugs based on imidazotriazine and pyrrolotriazine molecules. Further, it reacts with (1R:2R)-2-Hydroxy-πnanone-3 to prepare isopropyl (1R,2R,5R)-2′-[(2-hydroxypinan-3-ylene)amino]propanoate.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39825-33-7
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD08059709,MFCD08059709,MFCD12796134
InChI Key QDQVXVRZVCTVHE-UHFFFAOYNA-N
Synonym s-isopropyl 2-aminopropanoate, h-ala-oipr, l-alanine, 1-methylethyl ester, isopropyl 2s-2-aminopropanoate, alanine isopropyl ester, alanine, isopropyl ester, l-alanine isopropyl ester, propan-2-yl 2s-2-aminopropanoate
PubChem CID 13089460
IUPAC Name propan-2-yl 2-aminopropanoate
SMILES CC(C)OC(=O)C(C)N

Specifications

Quantity 1 g
Formula Weight 167.63
Percent Purity 95%
Chemical Name or Material L-Alanine isopropyl ester hydrochloride
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.