Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanine Isopropyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A258825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A258825G 25 g
A25885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A258825G Supplier TCI America Supplier No. A258825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62062-65-1
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD08059709
InChI Key YAQKNCSWDMGPOY-UHFFFAOYNA-N
Synonym (S)-2-Aminopropionic Acid Isopropyl Ester Hydrochloride, Isopropyl (S)-2-Aminopropionate Hydrochloride
PubChem CID 13089459
IUPAC Name propan-2-yl 2-aminopropanoate hydrochloride
SMILES Cl.CC(C)OC(=O)C(C)N

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 25 g
Formula Weight 167.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Alanine Isopropyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.