Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Alanyl-L-glutamine NA, Ajinomoto

Catalog No. 502946065
Encompass
Change view
Click to view available options
Quantity:
500 g
5 kg
20 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-294-6065 500 g
50-294-6074 20 kg
50-294-5993 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-294-6065 Supplier Thermo Scientific Production Chemicals and Services Supplier No. A32712
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 39537-23-0
Molecular Weight (g/mol) 217.22

Specifications

Packaging Poly Drum
Quantity 500 g
Percent Purity 97.0 to 103.0%
Chemical Name or Material L-Alanyl-L-Glutamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.