Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Arabinose Crystalline MP Biomedicals

Catalog No. ICN10070625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10070625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10070625 Supplier MP Biomedicals, Inc Supplier No. 0210070625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5328-37-0
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key PYMYPHUHKUWMLA-VAYJURFESA-N
Synonym l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
PubChem CID 5460291
ChEBI CHEBI:6182
IUPAC Name (2R,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Density 1.585g/mL (20°C)
Quantity 25 g
Formula Weight 150.1
Physical Form Crystalline
Chemical Name or Material L-(+)-Arabinose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.