Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine, 99%, [Free Base], MP Biomedicals

Catalog No. ICN19462690 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19462690 500 g
ICN19462625 25 g
ICN19462680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19462690 Supplier MP Biomedicals, Inc Supplier No. 0219462690
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 74-79-3 | C6H14N4O2 | 174.20 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Quantity 500 g
Formula Weight 174.2
Percent Purity 100%
Grade Cell Culture
Chemical Name or Material L-Arginine, 99%, Free Base
Recommended Storage Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.