Learn More
Description
Specifications
Specifications
| Melting Point | 223°C |
| pH | 11.4 |
| Color | White |
| Identification | Pass Test |
| Infrared Spectrum | Conforms to reference |
| Assay Percent Range | ≥99 % |
| Loss on Drying | 0.3% max. |
| Packaging | Poly Bottle |
| Specific Rotation | +26.3 to +27.7° (+ or -) |
| Merck Index | 15, 770 |
| Show More |
Safety and Handling
| Recommended Storage | RT |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.