Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine Hydrochloride, Multi-Compendial, U.S.P., J.T. Baker™

Catalog No. 02003175 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-003-175 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-175 Supplier Avantor Supplier No. 206706

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
PubChem CID 66250
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Packaging Poly Bottle
Quantity 1 kg
Formula Weight 210.66
Grade Multi-Compendial/USP
Specific Gravity 1L = 1.42kg
Chemical Name or Material L-Arginine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.