Learn More
Pfanstiehl Inc L-Arginine (Inhalation Grade) High Purity, Low Endotoxin, Low Metals, USP, EP, JP, ChP (A-170-IG) 100G GMP, Multi-compendial - CAS: 74-79-3 Injectable Excipient Grade
Supplier: Pfanstiehl Inc A170IGRD
Pfanstiehl L-Arginine, High Purity, Low Endotoxin, Low Metals is a multi-compendial amino acid that has been specifically designed and formulated for therapeutic inhalation formulations and to remove impurities such a endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl L-Arginine meets parenteral (injectable) excipient grade standards, making it ideal for pharmaceutical, biologic, and vaccine bioprocessing and formulation. Pfanstiehl L-Arginine is often used to protect and prevent aggregation of therapeutic proteins and is available as USP (United States Pharmacopoeia), EP (Ph. Eur.) (European Pharmacopoeia), JP (Japanese Pharmacopoeia) and ChP (Chinese Pharmacopoeia). Compendial, U.S.P. , cGMP ,
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.