Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-(+)-Ascorbic acid, Electrophoresis Grade, ≥99%

Click to view available options
Quantity:
50g
100g
50-81-7
C6H8O6
176.124
MFCD00064328
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
l-ascorbic acid,ascorbic acid,vitamin c,l-ascorbate,ascorbate,ascorbicap,l +-ascorbic acid,cevitamic acid,ascoltin,hybrin
54670067
CHEBI:29073
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
C(C(C1C(=C(C(=O)O1)O)O)O)O
50-81-7
C6H8O6
176.124
MFCD00064328
CIWBSHSKHKDKBQ-JLAZNSOCSA-N
l-ascorbic acid,ascorbic acid,vitamin c,l-ascorbate,ascorbate,ascorbicap,l +-ascorbic acid,cevitamic acid,ascoltin,hybrin
54670067
CHEBI:29073
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
C(C(C1C(=C(C(=O)O1)O)O)O)O
Quantity 50g
Density 1.65g/mL
Melting Point 190°C to 193°C (decomposition)
Color White
Beilstein 84272
Merck Index 14,830
Solubility Information Soluble in water at 0.5M
Formula Weight 176.12
Physical Form Powder
Percent Purity ≥99%
Grade Electrophoresis
Chemical Name or Material L-(+)-Ascorbic acid
Show More Show Less

RTECSNumber : CI7650000

TSCA : TSCA