Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Ascorbic Acid Sodium Salt, MP Biomedicals

Catalog No. ICN10289090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10289090 500 g
ICN10289025 25 g
ICN10289091 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10289090 Supplier MP Biomedicals, Inc Supplier No. 0210289090
Add to Cart
Add to Cart

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00082340
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
PubChem CID 131674100
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 500 g
Formula Weight 198.11
Physical Form Powder
Chemical Name or Material L-Ascorbic Acid Sodium Salt, Antimicrobial, Antioxidant, Reductant for the isolation of chloroplasts
EINECSNumber 205-126-1
storageNote1 Protect from light.
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.